Please login to your online account to display your discounted pricing

N1-(4-Butylphenyl)acetamide, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00828EA

 View more versions of this product

Catalog No. TL00828EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.27
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material N1-(4-Butylphenyl)acetamide
Assay Percent Range 97%
Molecular Formula C12H17NO
Linear Formula CH3CH2CH2CH2C6H4NHC(O)CH3
Packaging Amber Glass Bottle
MDL Number MFCD00277211
CAS 3663-20-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.

Specifications

Chemical Name or Material N1-(4-Butylphenyl)acetamide
CAS 3663-20-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H17NO
Linear Formula CH3CH2CH2CH2C6H4NHC(O)CH3
Formula Weight 191.27
MDL Number MFCD00277211
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.