Please login to your online account to display your discounted pricing

N1-(4-Aminophenyl)-N1-methylacetamide, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  BTB13402DA

 View more versions of this product

Catalog No. BTB13402DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 164.21
Quantity 1g
Assay Tech..
Chemical Name or Material N1-(4-Aminophenyl)-N1-methylacetamide
CAS 119-63-1
Molecular Formula C9H12N2O
Assay Percent Range 97%
Health Hazard 1 Warning

Specifications

Chemical Name or Material N1-(4-Aminophenyl)-N1-methylacetamide
Assay Tech..
Assay Percent Range 97%
Molecular Formula C9H12N2O
Formula Weight 164.21
Accessories

Warning

SDS & Safety Documents