Please login to your online account to display your discounted pricing

N1-(4-acetyl-3-hydroxyphenyl)acetamide, Maybridge

Chemical Identifiers

Chemical Name or Material N1-(4-Acetyl-3-hydroxyphenyl)acetamide
Assay Tech.
Molecular Formula C10H11NO3
Formula Weight 193.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB10058DE

 
fisher scientific company
BTB10058DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB10058DA

  SDS
fisher scientific company
BTB10058DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.2
Purity Grade Technical Grade
Quantity 5g
Assay Tech.
CAS 40547-58-8
Packaging Amber Glass Bottle
Molecular Formula C10H11NO3
Chemical Name or Material N1-(4-Acetyl-3-hydroxyphenyl)acetamide