Please login to your online account to display your discounted pricing

N1-(3-acetyl-4-hydroxyphenyl)acetamide, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  BTB10078DA

 View more versions of this product

Catalog No. BTB10078DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 193.2
Quantity 1g
Assay 97%
Molecular Formula C10H11NO3
CAS 7298-67-1
Assay Percent Range 97%
Chemical Name or Material N1-(3-Acetyl-4-hydroxyphenyl)acetamide

Specifications

Chemical Name or Material N1-(3-Acetyl-4-hydroxyphenyl)acetamide
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11NO3
Formula Weight 193.2