Please login to your online account to display your discounted pricing

N1-(2-Chlorophenyl)-2-bromoacetamide, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material N1-(2-chlorophenyl)-2-bromoacetamide
CAS 1292900
CAS Min % 97.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H7BrClNO
Linear Formula ClC6H4NHC(O)CH2Br
Formula Weight 248.51
MDL Number MFCD00173815
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

SPB05477EA

 
fisher scientific company
SPB05477EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.51
Quantity 10g
Assay 95%
CAS 1292900
CAS Min % 97.0
Chemical Name or Material N1-(2-chlorophenyl)-2-bromoacetamide
Molecular Formula C8H7BrClNO
Linear Formula ClC6H4NHC(O)CH2Br
MDL Number MFCD00173815
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.