Please login to your online account to display your discounted pricing

N-Vinylphthalimide, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC370200250

 View more versions of this product

Catalog No. AC370200250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 173.17
Quantity 25g
Color Off-white to light yellow
Melting Point 84° to 88°C
CAS Max % 100.0
Chemical Name or Material N-Vinylphthalimide
Assay Percent Range 99%
Molecular Formula C10H7NO2
MDL Number MFCD00078446
Health Hazard 1 Warning
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 3485-84-5
CAS Min % 98.5
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

Specifications

Chemical Name or Material N-Vinylphthalimide
CAS 3485-84-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00078446
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.

SDS & Safety Documents