Please login to your online account to display your discounted pricing

N-Vinylphthalimide, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Vinylphthalimide
CAS 3485-84-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00078446
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC370200250

  SDS
thermo fisher scientific
AC370200250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC370200050

  SDS
fisher scientific acros
AC370200050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 173.17
Physical Form Crystalline Powder
Quantity 25g
Color Off-white to light yellow
Melting Point 84° to 88°C
CAS Max % 100.0
MDL Number MFCD00078446
Molecular Formula C10H7NO2
Chemical Name or Material N-Vinylphthalimide
Assay Percent Range 99%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
CAS Min % 98.5
Packaging Glass bottle
CAS 3485-84-5
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.