Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Vinylphthalimide, 99%

$238.07 - $238.07

Chemical Identifiers

Chemical Name or Material N-Vinylphthalimide
CAS 3485-84-5
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00078446
Beilstein 127880
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0933506

  SDS
alfa aesar™
L0933506
5g Each for $238.07
Description & Specifications

Specifications

Formula Weight 173.17
Chemical Name or Material N-Vinylphthalimide
MDL Number MFCD00078446
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H7NO2
Assay 99%
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Odorless
Beilstein 127880
Health Hazard 1 Warning
Melting Point 84° to 88°C
Quantity 5g
CAS 3485-84-5
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.