Learn More
N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140925000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Vinyl-2-pyrrolidone | |
88-12-0 | |
100.0 | |
Colorless to Yellow | |
218°C | |
Authentic | |
Glass bottle | |
1.5119 to 1.5139 | |
500 mL | |
15,7814 | |
Solubility in water: 52.1g/L (25°C). Other solubilities: miscible with acetone,diethyl ether,ethanol,,toluene,benzene,very good soluble in organic solvents | |
C=CN1CCCC1=O | |
111.14 | |
6917 | |
CHEBI:82551 | |
99% |
98% | |
98.5 | |
13°C to 14°C | |
1.0430g/mL | |
95°C | |
0.2% max. (Pyrrolidone) (GC) | |
C6H9NO | |
MFCD00003197 | |
1.043 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
0.1% flaked caustic soda | |
2.4 mPa.s (20°C) | |
111.14 | |
Liquid |
Chemical Identifiers
88-12-0 | |
111.14 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
C6H9NO | |
MFCD00003197 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
CHEBI:82551 | |
C=CN1CCCC1=O |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 201-800-4
RTECSNumber : UY6107000
TSCA : TSCA
Recommended Storage : Product may darken on storage
RUO – Research Use Only