Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Undecyl methacrylate, 96%

$51.84 - $168.76

Chemical Identifiers

Chemical Name or Material n-Undecyl methacrylate
CAS 16493-35-9
Molecular Formula C15H28O2
Formula Weight 240.39
MDL Number MFCD00078348
Beilstein 2084688
Synonym Methacrylic acid n-undecyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1154406

  SDS
alfa aesar™
L1154406
5g Each for $51.84

AAL1154414

  SDS
alfa aesar™
L1154414
25g Each for $168.76
Description & Specifications

Specifications

Formula Weight 240.39
Chemical Name or Material n-Undecyl methacrylate
MDL Number MFCD00078348
Synonym Methacrylic acid n-undecyl ester
Health Hazard 1 Warning
CAS 16493-35-9
TSCA TSCA
Boiling Point 148°C (1mmHg)
Beilstein 2084688
Molecular Formula C15H28O2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.