missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Undecane, 99%

Manufacturer:  Alfa Aesar™ 3180718

 View more versions of this product

Catalog No. AA3180718



Specifications

Specifications

n-Undecane
60° (140°F)
1.4173
Colorless
50g
C11H24
MFCD00008959
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
CCCCCCCCCCC
156.313
CHEBI:46342
Liquid
0.74g/mL
Mild
196°C
-26°C
1120-21-4
CH3(CH2)9CH3
UN2330
RSJKGSCJYJTIGS-UHFFFAOYSA-N
undecane
14257
156.3
99%

Chemical Identifiers

1120-21-4
156.313
RSJKGSCJYJTIGS-UHFFFAOYSA-N
14257
undecane
C11H24
MFCD00008959
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
CHEBI:46342
CCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280f-P301+P330+P331-P315
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: UNDECANE

RTECSNumber : YQ1525000

TSCA : TSCA

SDS Documents