Learn More
n-Undecane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03180730
Description
n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Undecane | |
-26°C | |
0.74 g/mL | |
60°C (140°F) | |
C11H24 | |
CH3(CH2)9CH3 | |
250 g | |
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
undecane | |
14257 | |
156.3 | |
Liquid |
1120-21-4 | |
Colorless | |
196°C | |
Mild | |
1.4173 | |
MFCD00008959 | |
UN2330 | |
Miscible with ethyl alcohol and ether. Immiscible with water. | |
CCCCCCCCCCC | |
156.31 | |
CHEBI:46342 | |
99% |
Chemical Identifiers
1120-21-4 | |
156.31 | |
RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
14257 | |
undecane |
C11H24 | |
MFCD00008959 | |
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
CHEBI:46342 | |
CCCCCCCCCCC |
Safety and Handling
GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P280-P301+P310-P331-P370+P378q-P501c
H227-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: UNDECANE
EINECSNumber : 214-300-6
RTECSNumber : YQ1525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only