Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Tritylglycine ethyl ester, 98%

$201.91 - $711.19

Chemical Identifiers

Chemical Name or Material N-Tritylglycine ethyl ester
CAS 18514-46-0
Molecular Formula (C6H5)3CNHCH2CO2CH2CH3
Formula Weight 345.44
MDL Number MFCD00026893
Beilstein 3000902
Synonym N-(Triphenylmethyl)glycine ethyl ester; Trt-Gly-OEt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0828118

  SDS
alfa aesar™
L0828118
50g Each for $201.91

AAL0828130

  SDS
alfa aesar™
L0828130
250g Each for $711.19
Description & Specifications

Specifications

Formula Weight 345.44
Chemical Name or Material N-Tritylglycine ethyl ester
MDL Number MFCD00026893
Assay 98%
CAS 18514-46-0
Molecular Formula (C6H5)3CNHCH2CO2CH2CH3
Melting Point 112° to 116°C
Synonym N-(Triphenylmethyl)glycine ethyl ester; Trt-Gly-OEt
Beilstein 3000902
Quantity 50g