Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Tritylglycine, 97%

$104.42 - $313.41

Chemical Identifiers

Chemical Name or Material N-Tritylglycine
CAS 5893-05-0
Molecular Formula (C6H5)3CNHCH2CO2H
Formula Weight 317.39
MDL Number MFCD00004276
Beilstein 1998718
Synonym N-(Triphenylmethyl)glycine; Trt-Gly-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0909514

  SDS
alfa aesar™
L0909514
25g Each for $104.42

AAL0909522

  SDS
alfa aesar™
L0909522
100g Each for $313.41
Description & Specifications

Specifications

Formula Weight 317.39
MDL Number MFCD00004276
Molecular Formula (C6H5)3CNHCH2CO2H
CAS 5893-05-0
Synonym N-(Triphenylmethyl)glycine; Trt-Gly-OH
Assay 97%
Melting Point 170° to 174°C
Quantity 25g
Beilstein 1998718
Chemical Name or Material N-Tritylglycine