Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Trityl-L-alanine diethylammonium salt, 97%

Chemical Identifiers

Chemical Name or Material N-Trityl-L-alanine diethylammonium salt
CAS 80514-65-4
Formula Weight 404.55
MDL Number MFCD02094387
Beilstein 3923313
Synonym Trt-L-Ala.DEA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6692206

 
alfa aesar™
H6692206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692214

 
alfa aesar™
H6692214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692222

 
alfa aesar™
H6692222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 404.55
Chemical Name or Material N-Trityl-L-alanine diethylammonium salt
MDL Number MFCD02094387
CAS 80514-65-4
Physical Form Solid
Assay 97%
Synonym Trt-L-Ala.DEA
Beilstein 3923313
Quantity 5g