missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Triphenylmethyl)-L-serine Methyl Ester 98.0+%, TCI America™
Supplier: TCI America T287025G
Specifications
| N-(Triphenylmethyl)-L-serine Methyl Ester | |
| 148°C | |
| C23H23NO3 | |
| 25 g | |
| LXAWQKKSNNYYEK-NRFANRHFSA-N | |
| methyl (2S)-3-hydroxy-2-(tritylamino)propanoate | |
| 7128367 | |
| ≥98.0% (GC,T) |
| 4465-44-5 | |
| White-Yellow | |
| MFCD00192570 | |
| N-Trityl-L-serine Methyl Ester, Trt-Ser-OMe | |
| COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 361.441 | |
| 361.44 | |
| Crystalline Powder |
Chemical Identifiers
| 4465-44-5 | |
| 361.441 | |
| LXAWQKKSNNYYEK-NRFANRHFSA-N | |
| 7128367 | |
| COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
| C23H23NO3 | |
| MFCD00192570 | |
| N-Trityl-L-serine Methyl Ester, Trt-Ser-OMe | |
| methyl (2S)-3-hydroxy-2-(tritylamino)propanoate |
Safety and Handling
TSCA : No