missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Triphenylmethyl)-DL-serine Methyl Ester, TCI America™

Manufacturer:  TCI America T25005G

 View more versions of this product

Catalog No. T25005G



Specifications

Specifications

N-(Triphenylmethyl)-DL-serine Methyl Ester
146°C
13515-76-9
N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe
COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
361.441
361.44
≥98.0% (GC,T)
White
5g
C23H23NO3
LXAWQKKSNNYYEK-UHFFFAOYSA-N
methyl 3-hydroxy-2-(tritylamino)propanoate
279018
Crystal-Powder at 20°C

Chemical Identifiers

13515-76-9
361.441
N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe
methyl 3-hydroxy-2-(tritylamino)propanoate
C23H23NO3
LXAWQKKSNNYYEK-UHFFFAOYSA-N
279018
COC(=O)C(CO)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
Safety and Handling

Safety and Handling

TSCA : No

Documents