Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Trimethylsilylacetamide, 95%

$59.24 - $688.99

Chemical Identifiers

Chemical Name or Material N-Trimethylsilylacetamide
CAS 13435-12-6
Molecular Formula CH3C(O)NHSi(CH3)3
Formula Weight 131.25g/mol
MDL Number MFCD00008671
Beilstein 741928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1388414

 
alfa aesar™
A13884-14
25g Each for $59.24

AAA1388422

 
alfa aesar™
A13884-22
100g Each for $155.70

AAA1388436

 
alfa aesar™
A13884-36
500g Each for $688.99
Description & Specifications

Specifications

Melting Point 39° to 43°C
Boiling Point 104° to 106°C (35mmHg)
Quantity 25g
Flash Point 70°C (158°F)
Odor Odorless
Chemical Name or Material N-Trimethylsilylacetamide
CAS 13435-12-6
Assay 95%
Molecular Formula CH3C(O)NHSi(CH3)3
Formula Weight 131.25g/mol
MDL Number MFCD00008671
Beilstein 741928
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AD0250000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.