Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[(Trimethylsilyl)methyl]benzylamine, 95%

Chemical Identifiers

Chemical Name or Material N-[(Trimethylsilyl)methyl]benzylamine
CAS 53215-95-5
Formula Weight 193.36
MDL Number MFCD00012192
Beilstein 2802488
Synonym N-Benzyl-1-(trimethylsilyl)methylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6445006

  SDS
alfa aesar™
H6445006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6445022

  SDS
alfa aesar™
H6445022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6445014

  SDS
alfa aesar™
H6445014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.36
Boiling Point 89° to 90°C (5mmHg)
Assay 95%
Color Pale yellow
CAS 53215-95-5
Synonym N-Benzyl-1-(trimethylsilyl)methylamine
Refractive Index 1.4960
Physical Form Liquid
MDL Number MFCD00012192
Beilstein 2802488
Chemical Name or Material N-[(Trimethylsilyl)methyl]benzylamine
Quantity 5g
Density 0.88g/mL