Learn More
N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, ACROS Organics, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 93 deg.C to 94 deg.C (13.0mmHg), Colorless to Yellow, Melting Point: -42 deg.C, Molecular Weight: 140.26, Percent Purity: 97%, Assay Percent Range: 96% min. (GC), CAS: 18156-74-6
Supplier: Thermo Scientific Chemicals 167130250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentSpecifications
N-(Trimethylsilyl)imidazole | |
96.0 | |
-42°C | |
0.9500g/mL | |
5°C | |
96% min. (GC) | |
C6H12N2Si | |
MFCD00005280 | |
07,399; 11,575; 01,337; 02,177 | |
n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl | |
YKFRUJSEPGHZFJ-UHFFFAOYSA-N | |
imidazol-1-yl(trimethyl)silane | |
28925 | |
CHEBI:85063 | |
97% |
18156-74-6 | |
100.0 | |
Colorless to Yellow | |
93°C to 94°C (13.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4750 to 1.478 | |
25 g | |
0.95 | |
Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents | |
C[Si](C)(C)N1C=CN=C1 | |
140.26 | |
2.5 mm2/s (25°C) | |
140.26 | |
Liquid |
Chemical Identifiers
18156-74-6 | |
140.26 | |
YKFRUJSEPGHZFJ-UHFFFAOYSA-N | |
28925 | |
imidazol-1-yl(trimethyl)silane |
C6H12N2Si | |
MFCD00005280 | |
n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl | |
CHEBI:85063 | |
C[Si](C)(C)N1C=CN=C1 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 242-040-3
RUO – Research Use Only