Learn More
N-(Trimethylsilyl)diethylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1814914
Description
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
125°C to 126°C | |
-10°C | |
25 g | |
MFCD00009040 | |
635718 | |
n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine | |
JOOMLFKONHCLCJ-UHFFFAOYSA-N | |
N-ethyl-N-trimethylsilylethanamine | |
70454 | |
145.32 | |
Amine-like | |
Moisture sensitive |
N-(Trimethylsilyl)diethylamine | |
996-50-9 | |
C7H19NSi | |
UN2733 | |
10°C (50°F) | |
Hydrolyzes in water. | |
CCN(CC)[Si](C)(C)C | |
145.32 | |
CHEBI:85070 | |
98% | |
1.411 | |
0.76 |
Chemical Identifiers
996-50-9 | |
145.32 | |
JOOMLFKONHCLCJ-UHFFFAOYSA-N | |
70454 | |
N-ethyl-N-trimethylsilylethanamine |
C7H19NSi | |
MFCD00009040 | |
n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine | |
CHEBI:85070 | |
CCN(CC)[Si](C)(C)C |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 213-637-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only