missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Trimethylsilyl)acetamide, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC207591000

 View more versions of this product

Catalog No. AC207591000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(Trimethylsilyl)acetamide
Glass bottle
40°C to 46°C
13435-12-6
100.0
C5H13NOSi
MFCD00008671
n-trimethylsilyl acetamide, trimethylsilylacetamide, acetamide, n-trimethylsilyl, tms acetamide, acetylamino trimethylsilane, trimethylsilyl acetamide, acetamide, tms derivative, n-trimethylsilyl-acetamide, n-tms-acetamide, n-trimethylsilylacetoamide
CC(=O)N[Si](C)(C)C
131.25
131.25
≥96.0%
Authentic
White to Yellow
100g
96.0
97%
CH3CONHSi(CH3)3
01,1235; 16,178
LWFWUJCJKPUZLV-UHFFFAOYSA-N
N-trimethylsilylacetamide
25989
Crystalline Mass

Chemical Identifiers

13435-12-6
131.25
LWFWUJCJKPUZLV-UHFFFAOYSA-N
25989
CC(=O)N[Si](C)(C)C
C5H13NOSi
MFCD00008671
n-trimethylsilyl acetamide, trimethylsilylacetamide, acetamide, n-trimethylsilyl, tms acetamide, acetylamino trimethylsilane, trimethylsilyl acetamide, acetamide, tms derivative, n-trimethylsilyl-acetamide, n-tms-acetamide, n-trimethylsilylacetoamide
N-trimethylsilylacetamide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents