Please login to your online account to display your discounted pricing

N-(Trimethylsilyl)acetamide, 97%, ACROS Organics™

$76.36 - $76.36

Chemical Identifiers

Chemical Name or Material N-(Trimethylsilyl)acetamide
CAS 13435-12-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H13NOSi
Linear Formula CH3CONHSi(CH3)3
Formula Weight 131.25
MDL Number MFCD00008671
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC207591000

  SDS
thermo fisher scientific
AC207591000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC207590250

  SDS
thermo fisher scientific
207590250
Glass bottle 25g Each for $76.36
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Mass
Formula Weight 131.25
Quantity 100g
Color White to yellow
Melting Point 40° to 46°C
Fieser 01,1235; 16,178
CAS Min % 96.0
CAS Max % 100.0
Linear Formula CH3CONHSi(CH3)3
Packaging Glass bottle
Molecular Formula C5H13NOSi
CAS 13435-12-6
Assay Percent Range 97%
Chemical Name or Material N-(Trimethylsilyl)acetamide
MDL Number MFCD00008671
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.