Please login to your online account to display your discounted pricing

N-(α,α,α-Trifluoro-p-tolyl)piperazine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC367120010

 View more versions of this product

Catalog No. AC367120010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals
Formula Weight 230.23
Quantity 1g
Color Colorless to light yellow
Melting Point 85° to 90°C
CAS Max % 100.0
Name Note 98%
CAS 30459-17-7
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 1-(4-Trifluoromethylphenyl)piperazine
Chemical Name or Material N-(α, α, α-Trifluoro-p-tolyl)piperazine
Assay Percent Range 98%
Molecular Formula C11H13F3N3
MDL Number MFCD00040765
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-(α, α, α-Trifluoro-p-tolyl)piperazine
Name Note 98%
CAS 30459-17-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H13F3N3
Formula Weight 230.23
MDL Number MFCD00040765
Synonym 1-(4-Trifluoromethylphenyl)piperazine
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

SDS & Safety Documents