Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Tridecylbenzene, 97%

$56.04 - $56.04

Chemical Identifiers

Chemical Name or Material n-Tridecylbenzene
CAS 123-02-4
Molecular Formula C19H32
Formula Weight 260.47
MDL Number MFCD00008980
Beilstein 1947201
Merck Index 14,9659
Synonym 1-Phenyltridecane; Tridane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0196206

 
alfa aesar™
L0196206
5g Each for $56.04
Description & Specifications

Specifications

Formula Weight 260.47
Chemical Name or Material n-Tridecylbenzene
MDL Number MFCD00008980
TSCA TSCA
Assay 97%
Molecular Formula C19H32
CAS 123-02-4
Beilstein 1947201
Merck Index 14,9659
Synonym 1-Phenyltridecane; Tridane
Flash Point 112°C (233°F)
Boiling Point 340°C
Melting Point 10°C
Quantity 5g