Please login to your online account to display your discounted pricing

N-Tosylpyrrole, ≥99%, Acros Organics


Manufacturer: thermo fisher scientific  270060010

 View more versions of this product

Catalog No. AC270060010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 221.28
Physical Form Crystalline Powder
Quantity 1g
Color Beige to light brown
Melting Point 98° to 102°C
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00145014
CAS 17639-64-4
Name Note 99+%
Molecular Formula C11H11NO2S
Chemical Name or Material N-Tosylpyrrole
Assay Percent Range 98%

Specifications

Chemical Name or Material N-Tosylpyrrole
CAS 17639-64-4
CAS Min % 99.0
CAS Max % 100.0
Name Note 99+%
Molecular Formula C11H11NO2S
Formula Weight 221.28
MDL Number MFCD00145014