missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Tosylpyrrole, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC270060250
Specifications
N-Tosylpyrrole | |
Beige to Brown | |
25g | |
17639-64-4 | |
100.0 | |
C11H11NO2S | |
1-4-methylbenzenesulfonyl pyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-p-toluenesulfonyl pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-1h-pyrrole, 1-tosyl-pyrrole, 1-tosylpyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl, n-tosylpyrrole | |
CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2 | |
221.28 | |
221.28 | |
98% |
Authentic | |
103.0°C to 104.5°C | |
99+% | |
99.0 | |
97.5% min. (GC) | |
MFCD00145014 | |
OXWIEWFMRVJGNY-UHFFFAOYSA-N | |
1-(4-methylphenyl)sulfonylpyrrole | |
640087 | |
Crystalline Powder |
Chemical Identifiers
17639-64-4 | |
221.28 | |
OXWIEWFMRVJGNY-UHFFFAOYSA-N | |
640087 | |
CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2 |
C11H11NO2S | |
MFCD00145014 | |
1-4-methylbenzenesulfonyl pyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-p-toluenesulfonyl pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-1h-pyrrole, 1-tosyl-pyrrole, 1-tosylpyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl, n-tosylpyrrole | |
1-(4-methylphenyl)sulfonylpyrrole |
Safety and Handling
GHS Signal Word: Warning