missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Tosylpyrrole, 98%, ACROS Organics™

Chemical Identifiers

CAS 17639-64-4
Molecular Formula C11H11NO2S
Molecular Weight (g/mol) 221.28
MDL Number MFCD00145014
InChI Key OXWIEWFMRVJGNY-UHFFFAOYSA-N
Synonym 1-4-methylbenzenesulfonyl pyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-p-toluenesulfonyl pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-1h-pyrrole, 1-tosyl-pyrrole, 1-tosylpyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl, n-tosylpyrrole
PubChem CID 640087
IUPAC Name 1-(4-methylphenyl)sulfonylpyrrole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC270060250
SDS View Documents
Thermo Fisher Scientific
AC270060250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

17639-64-4
221.28
OXWIEWFMRVJGNY-UHFFFAOYSA-N
640087
CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2
C11H11NO2S
MFCD00145014
1-4-methylbenzenesulfonyl pyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-p-toluenesulfonyl pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-1h-pyrrole, 1-tosyl-pyrrole, 1-tosylpyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl, n-tosylpyrrole
1-(4-methylphenyl)sulfonylpyrrole

Specifications

Authentic
103.0°C to 104.5°C
17639-64-4
100.0
C11H11NO2S
1-4-methylbenzenesulfonyl pyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-p-toluenesulfonyl pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-1h-pyrrole, 1-tosyl-pyrrole, 1-tosylpyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl, n-tosylpyrrole
CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2
221.28
221.28
98%
Beige to Brown
25g
99.0
97.5% min. (GC)
MFCD00145014
OXWIEWFMRVJGNY-UHFFFAOYSA-N
1-(4-methylphenyl)sulfonylpyrrole
640087
Crystalline Powder
N-Tosylpyrrole, 99+%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS