Learn More
N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC441020050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-TIPS pyrrole-3-boronic acid pinacol ester | |
94.0 | |
White to Yellow | |
94% min. (GC) | |
C19H36BNO2Si | |
Glass bottle | |
5g | |
CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1 | |
349.40 | |
349.4 | |
95% |
365564-11-0 | |
100.0 | |
70°C to 72°C | |
Authentic | |
MFCD08063117 | |
1-triisopropylsilyl-1h-pyrrol-3-ylboronic acid pinacol ester, 1-triisopropylsilyl-1h-pyrrole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl pyrrole, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-pyrrole, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris 1-methylethyl silyl-1h-pyrrole, 3-pinacolateboryl-1-tips-pyrrole, 3-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris propan-2-yl silyl-1h-pyrrole, n-tips pyrrole-3-boronic acid pinacol ester, pubchem18460, pyrrole-3-boronic acid, pinacol ester; | |
GWFIZBYDIHGZRJ-UHFFFAOYSA-N | |
tri(propan-2-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrol-1-yl]silane | |
11450924 | |
Powder or Solid |
Chemical Identifiers
365564-11-0 | |
349.40 | |
GWFIZBYDIHGZRJ-UHFFFAOYSA-N | |
11450924 | |
CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1 |
C19H36BNO2Si | |
MFCD08063117 | |
1-triisopropylsilyl-1h-pyrrol-3-ylboronic acid pinacol ester, 1-triisopropylsilyl-1h-pyrrole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl pyrrole, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-pyrrole, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris 1-methylethyl silyl-1h-pyrrole, 3-pinacolateboryl-1-tips-pyrrole, 3-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris propan-2-yl silyl-1h-pyrrole, n-tips pyrrole-3-boronic acid pinacol ester, pubchem18460, pyrrole-3-boronic acid, pinacol ester; | |
tri(propan-2-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrol-1-yl]silane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only