Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Thionylaniline, 97%

Chemical Identifiers

Chemical Name or Material N-Thionylaniline
CAS 1122-83-4
Formula Weight 139.18
MDL Number MFCD00002084
Beilstein 507682
Synonym N-Sulfinylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5945114

 
alfa aesar™
H5945114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.18
Chemical Name or Material N-Thionylaniline
TSCA TSCA
Refractive Index 1.6275
Physical Form Liquid
Assay 97%
MDL Number MFCD00002084
CAS 1122-83-4
Beilstein 507682
Synonym N-Sulfinylaniline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H335-H227-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P285-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.236g/mL
Boiling Point 200°C
Color Pale brown
Quantity 25g

Applications
It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.

Safety and Handling

Danger

GHS P Statement
P210-P285-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H335-H227-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.