Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Tetradecyltrichlorosilane, 97%


Quantity 5g

Manufacturer: alfa aesar™  L0549506

 View more versions of this product

Catalog No. AAL0549506

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 331.83
Chemical Name or Material n-Tetradecyltrichlorosilane
TSCA TSCA
Assay 97%
Molecular Formula CH3(CH2)13SiCl3
MDL Number MFCD00051440
CAS 18402-22-7
UN Number UN2987
Beilstein 1764981
Synonym Myristyltrichlorosilane; Trichloro-n-tetradecylsilane
Boiling Point 155° to 157°C (3mmHg)
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

Specifications

Chemical Name or Material n-Tetradecyltrichlorosilane
CAS 18402-22-7
Molecular Formula CH3(CH2)13SiCl3
Formula Weight 331.83
MDL Number MFCD00051440
UN Number UN2987
Beilstein 1764981
Synonym Myristyltrichlorosilane; Trichloro-n-tetradecylsilane
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents