missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Tetradecane, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 3080914

 View more versions of this product

Catalog No. AA3080914



Specifications

Specifications

n-Tetradecane
25g
>99%
MFCD00008986
BGHCVCJVXZWKCC-UHFFFAOYSA-N
tetradecane
12389
198.39
0.763g/mL
629-59-4
C14H30
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
CCCCCCCCCCCCCC
198.394
CHEBI:41253
Liquid

Chemical Identifiers

629-59-4
198.394
BGHCVCJVXZWKCC-UHFFFAOYSA-N
12389
tetradecane
C14H30
MFCD00008986
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
CHEBI:41253
CCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

EINECSNumber : 211-096-0

SDS Documents