Learn More
n-Tetradecane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1467236
Description
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
n-Tetradecane | |
6°C | |
252°C to 254°C | |
99% | |
1.429 | |
MFCD00008986 | |
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine | |
BGHCVCJVXZWKCC-UHFFFAOYSA-N | |
tetradecane | |
12389 | |
198.39 | |
Liquid |
629-59-4 | |
0.7628 | |
99°C (211°F) | |
C14H30 | |
CH3(CH2)12CH3 | |
500 g | |
Soluble in ether,acetone and alcohol. Insoluble in water. | |
CCCCCCCCCCCCCC | |
198.394 | |
CHEBI:41253 | |
99% |
Chemical Identifiers
629-59-4 | |
198.394 | |
BGHCVCJVXZWKCC-UHFFFAOYSA-N | |
12389 | |
tetradecane |
C14H30 | |
MFCD00008986 | |
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine | |
CHEBI:41253 | |
CCCCCCCCCCCCCC |
Safety and Handling
GHS H Statement
H304-H336
P301+P310-P331-P501c
H304
EINECSNumber : 211-096-0
RTECSNumber : XB8000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only