missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Tetradecane, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1467222

 View more versions of this product

Catalog No. AAA1467222



Description

Description

Applications
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

n-Tetradecane
100g
99%
CH3(CH2)12CH3
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
CCCCCCCCCCCCCC
198.394
CHEBI:41253
Liquid
0.7628
629-59-4
C14H30
MFCD00008986
BGHCVCJVXZWKCC-UHFFFAOYSA-N
tetradecane
12389
198.39
99%

Chemical Identifiers

629-59-4
198.394
BGHCVCJVXZWKCC-UHFFFAOYSA-N
12389
tetradecane
C14H30
MFCD00008986
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
CHEBI:41253
CCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H304-H336

GHS P Statement
P261-P271-P301+P310-P304+P340-P312-P331-P405-P403+P233-P501a

Danger

RTECSNumber : XB8000000

TSCA : TSCA

Documents