missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-tert-Butylglycine hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H6427003

 View more versions of this product

Catalog No. AAH6427003



Specifications

Specifications

N-tert-Butylglycine hydrochloride
C6H14ClNO2
MFCD03788441
2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
CC(C)(C)NCC(=O)O.Cl
167.633
167.63
97%
223°C to 224°C
6939-23-7
(CH3)3CNHCH2COOH·HCl
6296616
QORBVLNEGUIPND-UHFFFAOYSA-N
2-(tert-butylamino)acetic acid;hydrochloride
17748195
Solid
White
1g

Chemical Identifiers

6939-23-7
167.633
QORBVLNEGUIPND-UHFFFAOYSA-N
17748195
CC(C)(C)NCC(=O)O.Cl
C6H14ClNO2
MFCD03788441
2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
2-(tert-butylamino)acetic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents