missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-tert-Butylglycine hydrochloride, 97%, Thermo Scientific Chemicals

$44.92 - $536.89

Chemical Identifiers

CAS 6939-23-7
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD03788441
InChI Key QORBVLNEGUIPND-UHFFFAOYSA-N
Synonym 2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
PubChem CID 17748195
IUPAC Name hydrogen 2-(tert-butylamino)acetic acid chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6427003
View Documents Promotion Details
Thermo Scientific Chemicals
H6427003
1 g
Each for $44.92
 
AAH6427006
View Documents Promotion Details
Thermo Scientific Chemicals
H6427006
5 g
Each for $137.97
 
AAH6427014
View Documents Promotion Details
Thermo Scientific Chemicals
H6427014
25 g
Each for $536.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6939-23-7
167.63
QORBVLNEGUIPND-UHFFFAOYSA-N
17748195
[H+].[Cl-].CC(C)(C)NCC(O)=O
C6H14ClNO2
MFCD03788441
2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
hydrogen 2-(tert-butylamino)acetic acid chloride

Specifications

223°C to 224°C
White
(CH3)3CNHCH2COOH·HCl
6296616
QORBVLNEGUIPND-UHFFFAOYSA-N
hydrogen 2-(tert-butylamino)acetic acid chloride
17748195
97%
Solid
6939-23-7
C6H14ClNO2
MFCD03788441
2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
[H+].[Cl-].CC(C)(C)NCC(O)=O
167.63
167.63
1 g
N-tert-Butylglycine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only