missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-tert-Butylcyclohexylamine, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC439830050

 View more versions of this product

Catalog No. AC439830050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-tert-Butylcyclohexylamine
96.0
0.8340g/mL
172°C to 174°C
96% min. (GC)
C10H21N
n-1,1-dimethylethyl cyclohexanamine, n-t-butylcyclohexylamine, n-tert-butyl cyclohexanamine, n-tert-butyl-n-cyclohexyl amine, n-tert-butylcyclohexylamine, n-tertbutyl-n-cyclohexyl amine, tert-butyl cyclohexylamine, tert-butyl-cyclohexylamine, tert-butylcyclohexyl amine, tert.-butyl-cyclohexyl-amine
MTEWAFVECQBILW-UHFFFAOYSA-N
CC(C)(C)NC1CCCCC1
155.29
155.29
97%
51609-06-4
100.0
Colorless to Yellow
50°C
Authentic
Glass bottle
5g
0.834
N-tert-butylcyclohexanamine
103920
Liquid

Chemical Identifiers

51609-06-4
155.29
n-1,1-dimethylethyl cyclohexanamine, n-t-butylcyclohexylamine, n-tert-butyl cyclohexanamine, n-tert-butyl-n-cyclohexyl amine, n-tert-butylcyclohexylamine, n-tertbutyl-n-cyclohexyl amine, tert-butyl cyclohexylamine, tert-butyl-cyclohexylamine, tert-butylcyclohexyl amine, tert.-butyl-cyclohexyl-amine
N-tert-butylcyclohexanamine
C10H21N
MTEWAFVECQBILW-UHFFFAOYSA-N
103920
CC(C)(C)NC1CCCCC1
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 257-320-

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only