Please login to your online account to display your discounted pricing

Alfa Aesar™ N-tert-Butylbenzamide, 98+%

$130.47 - $130.47

Chemical Identifiers

Chemical Name or Material N-tert-Butylbenzamide
CAS 5894-65-5
Molecular Formula C11H15NO
Formula Weight 177.25
MDL Number MFCD00041337
Beilstein 2042391
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0850914

  SDS
alfa aesar™
L0850914
25g Each for $130.47
Description & Specifications

Specifications

Formula Weight 177.25
Chemical Name or Material N-tert-Butylbenzamide
Assay ≥98%
Molecular Formula C11H15NO
CAS 5894-65-5
MDL Number MFCD00041337
Beilstein 2042391
Melting Point 133° to 135°C
Quantity 25g