Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(tert-Butyl)thiourea, 97%

$88.05 - $88.05

Chemical Identifiers

Chemical Name or Material N-(tert-Butyl)thiourea
CAS 7204-48-0
Molecular Formula C5H12N2S
Formula Weight 132.23
MDL Number MFCD00041192
Beilstein 1744506
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1197903

 
alfa aesar™
L1197903
1g Each for $88.05
Description & Specifications

Specifications

Formula Weight 132.23
Chemical Name or Material N-(tert-Butyl)thiourea
Assay 97%
Molecular Formula C5H12N2S
MDL Number MFCD00041192
CAS 7204-48-0
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point 172° to 176°C
Beilstein 1744506
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.