missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(tert-Butyl)thiourea, 97%

Manufacturer:  Alfa Aesar™ L1197906

 View more versions of this product

Catalog No. AAL1197906



Specifications

Specifications

N-(tert-Butyl)thiourea
172°C to 176°C
7204-48-0
MFCD00041192
1-tert-butyl-2-thiourea, 1-tert-butylthiourea, n-tert-butyl thiourea, tert-butyl-thiourea, 1-tert-butyl thiourea, t-butylthiourea, n-tertbutylthiourea, tert-butyl thiourea, n-t-butyl thiourea, acmc-1bb93
CC(C)(C)NC(=S)N
132.225
132.23
Odorless
5g
C5H12N2S
1744506
RYOCWONLFFPYMN-UHFFFAOYSA-N
tert-butylthiourea
737374
97%

Chemical Identifiers

7204-48-0
132.225
RYOCWONLFFPYMN-UHFFFAOYSA-N
737374
CC(C)(C)NC(=S)N
C5H12N2S
MFCD00041192
1-tert-butyl-2-thiourea, 1-tert-butylthiourea, n-tert-butyl thiourea, tert-butyl-thiourea, 1-tert-butyl thiourea, t-butylthiourea, n-tertbutylthiourea, tert-butyl thiourea, n-t-butyl thiourea, acmc-1bb93
tert-butylthiourea
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents