missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-tert-Butyl-4-nitrobenzenesulfonamide, 97%

$113.35 - $422.54

Chemical Identifiers

CAS 49690-09-7
Molecular Formula C10H14N2O4S
Molecular Weight (g/mol) 258.292
MDL Number MFCD00443679
InChI Key YVSUQTZCHSBFEN-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-1,1-dimethylethyl-4-nitro
PubChem CID 796974
IUPAC Name N-tert-butyl-4-nitrobenzenesulfonamide
SMILES CC(C)(C)NS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH58068MC View Documents Alfa Aesar™
H58068MC
100mg Each for $113.35
 
AAH58068ME View Documents Alfa Aesar™
H58068ME
500mg Each for $422.54
 
Specifications

Chemical Identifiers

49690-09-7
258.292
YVSUQTZCHSBFEN-UHFFFAOYSA-N
796974
CC(C)(C)NS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C10H14N2O4S
MFCD00443679
benzenesulfonamide, n-1,1-dimethylethyl-4-nitro
N-tert-butyl-4-nitrobenzenesulfonamide

Specifications

100mg
C10H14N2O4S
benzenesulfonamide, n-1,1-dimethylethyl-4-nitro
CC(C)(C)NS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
258.292
258.29
N-tert-Butyl-4-nitrobenzenesulfonamide
49690-09-7
MFCD00443679
YVSUQTZCHSBFEN-UHFFFAOYSA-N
N-tert-butyl-4-nitrobenzenesulfonamide
796974
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning