Please login to your online account to display your discounted pricing

Alfa Aesar™ N-tert-Butyl-3,5-dimethylaniline, 97%

Chemical Identifiers

Chemical Name or Material N-tert-Butyl-3,5-dimethylaniline
CAS 110993-40-3
Formula Weight 177.29
MDL Number MFCD07440135
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5516403

 
alfa aesar™
H5516403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5516406

 
alfa aesar™
H5516406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55164MD

 
alfa aesar™
H55164MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.29
Boiling Point 69° to 70°C (0.75mmHg)
Chemical Name or Material N-tert-Butyl-3,5-dimethylaniline
Refractive Index 1.5190
Physical Form Liquid
Assay 97%
MDL Number MFCD07440135
CAS 110993-40-3
Density 0.901g/mL
Color Colorless to brown
Quantity 1g