missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(tert-Butoxycarbonyloxy)succinimide, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436240250
Specifications
N-(tert-Butoxycarbonyloxy)succinimide | |
Authentic | |
White | |
25g | |
97.5 | |
97% | |
1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, boc-osu, boc-osu, tert-butyl n-succinimidyl carbonate, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, n-tert-butoxycarbonyloxy succinimide, n-tert-butoxycarbonyloxy succinimide boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, tert-butyl succinimido carbonate | |
CC(C)(C)OC(=O)ON1C(=O)CCC1=O | |
215.21 | |
215.21 | |
97% |
48.22 to 52.24% C, 6.25 to 6.77% N | |
Glass Bottle | |
88°C to 91°C | |
13139-12-3 | |
100.0 | |
C9H13NO5 | |
VTGFSVGZCYYHLO-UHFFFAOYSA-N | |
tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate | |
83168 | |
Powder |
Chemical Identifiers
13139-12-3 | |
215.21 | |
1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, boc-osu, boc-osu, tert-butyl n-succinimidyl carbonate, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, n-tert-butoxycarbonyloxy succinimide, n-tert-butoxycarbonyloxy succinimide boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, tert-butyl succinimido carbonate | |
tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate |
C9H13NO5 | |
VTGFSVGZCYYHLO-UHFFFAOYSA-N | |
83168 | |
CC(C)(C)OC(=O)ON1C(=O)CCC1=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 236-071-1