missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(tert-Butoxycarbonyloxy)succinimide, 98+%

Manufacturer:  Alfa Aesar™ H5449303

 View more versions of this product

Catalog No. AAH5449303



Specifications

Specifications

N-(tert-Butoxycarbonyloxy)succinimide
∽95°C (decomposition)
13139-12-3
MFCD00037903
VTGFSVGZCYYHLO-UHFFFAOYSA-N
tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
83168
≥98%
Moisture sensitive
1g
C9H13NO5
1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione
CC(C)(C)OC(=O)ON1C(=O)CCC1=O
215.205
215.2

Chemical Identifiers

13139-12-3
215.205
VTGFSVGZCYYHLO-UHFFFAOYSA-N
83168
CC(C)(C)OC(=O)ON1C(=O)CCC1=O
C9H13NO5
MFCD00037903
1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione
tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

EINECSNumber : 236-071-1

TSCA : Yes

Recommended Storage : Store at -20°C; Store under Nitrogen

Documents