missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline Methyl Ester, TCI America™

$99.31 - $293.34

Chemical Identifiers

CAS 88043-21-4
Molecular Formula C18H25NO7S
Molecular Weight (g/mol) 399.458
MDL Number MFCD02179383
InChI Key UKVKNGDXVREPDE-HIFRSBDPSA-N
Synonym boc-trans-4-tosyloxy-l-proline methyl ester, n-boc-trans-4-tosyloxy-l-proline methyl ester, 4r-4-tso-n-boc-pro-ome, n-boc-trans-4-p-tosyloxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-tosyloxy pyrrolidine-1,2-dicarboxylate, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-4-methylbenzenesulfonyl oxy pyrrolidine-1,2-dicarboxylate, ksc659e9j, boc-trans-4-tosyloxy-l-prolinemethylester
PubChem CID 13425772
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-(4-methylphenyl)sulfonyloxypyrrolidine-1,2-dicarboxylate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CC(N(C2)C(=O)OC(C)(C)C)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B5247200MG View Documents TCI America
B5247200MG
200mg Each for $99.31
 
B52471G View Documents TCI America
B52471G
1g Each for $293.34
 
Specifications

Chemical Identifiers

88043-21-4
399.458
UKVKNGDXVREPDE-HIFRSBDPSA-N
13425772
CC1=CC=C(C=C1)S(=O)(=O)OC2CC(N(C2)C(=O)OC(C)(C)C)C(=O)OC
C18H25NO7S
MFCD02179383
boc-trans-4-tosyloxy-l-proline methyl ester, n-boc-trans-4-tosyloxy-l-proline methyl ester, 4r-4-tso-n-boc-pro-ome, n-boc-trans-4-p-tosyloxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-tosyloxy pyrrolidine-1,2-dicarboxylate, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-4-methylbenzenesulfonyl oxy pyrrolidine-1,2-dicarboxylate, ksc659e9j, boc-trans-4-tosyloxy-l-prolinemethylester
1-O-tert-butyl 2-O-methyl (2S,4R)-4-(4-methylphenyl)sulfonyloxypyrrolidine-1,2-dicarboxylate

Specifications

White-Yellow
200mg
C18H25NO7S
boc-trans-4-tosyloxy-l-proline methyl ester, n-boc-trans-4-tosyloxy-l-proline methyl ester, 4r-4-tso-n-boc-pro-ome, n-boc-trans-4-p-tosyloxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-tosyloxy pyrrolidine-1,2-dicarboxylate, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-4-methylbenzenesulfonyl oxy pyrrolidine-1,2-dicarboxylate, ksc659e9j, boc-trans-4-tosyloxy-l-prolinemethylester
CC1=CC=C(C=C1)S(=O)(=O)OC2CC(N(C2)C(=O)OC(C)(C)C)C(=O)OC
399.458
399.46
≥98.0% (HPLC,N)
79°C
88043-21-4
MFCD02179383
UKVKNGDXVREPDE-HIFRSBDPSA-N
1-O-tert-butyl 2-O-methyl (2S,4R)-4-(4-methylphenyl)sulfonyloxypyrrolidine-1,2-dicarboxylate
13425772
Crystal-Powder at 20°C
N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline Methyl Ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator