missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-O-benzyl-L-serine N-Succinimidyl Ester, TCI America™

$45.83 - $137.50

Chemical Identifiers

CAS 13650-73-2
Molecular Formula C19H24N2O7
Molecular Weight (g/mol) 392.408
MDL Number MFCD00037908
InChI Key NTFYOALQRQBBDG-AWEZNQCLSA-N
Synonym boc-ser bzl-osu, n-tert-butoxycarbonyl-o-benzyl-l-serine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, 2,5-dioxopyrrolidin-1-yl 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, boc-ser bzl osu, n-boc-3-benzyloxy-ala-succinimidyl oh, boc-ser bzl-osu hplc, n-boc-o-benzyl-l-serine n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl o-benzyl-n-t-butoxycarbonyl-l-serinate
PubChem CID 12879025
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoate
SMILES CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22051G View Documents TCI America
B22051G
1g Each for $45.83
 
B22055G View Documents TCI America
B22055G
5g Each for $137.50
 
Specifications

Chemical Identifiers

13650-73-2
392.408
NTFYOALQRQBBDG-AWEZNQCLSA-N
12879025
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
C19H24N2O7
MFCD00037908
boc-ser bzl-osu, n-tert-butoxycarbonyl-o-benzyl-l-serine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, 2,5-dioxopyrrolidin-1-yl 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, boc-ser bzl osu, n-boc-3-benzyloxy-ala-succinimidyl oh, boc-ser bzl-osu hplc, n-boc-o-benzyl-l-serine n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl o-benzyl-n-t-butoxycarbonyl-l-serinate
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoate

Specifications

Undesignated
1g
C19H24N2O7
boc-ser bzl-osu, n-tert-butoxycarbonyl-o-benzyl-l-serine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, 2,5-dioxopyrrolidin-1-yl 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, boc-ser bzl osu, n-boc-3-benzyloxy-ala-succinimidyl oh, boc-ser bzl-osu hplc, n-boc-o-benzyl-l-serine n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl o-benzyl-n-t-butoxycarbonyl-l-serinate
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
392.408
392.41
≥98.0% (N)
108°C
13650-73-2
MFCD00037908
NTFYOALQRQBBDG-AWEZNQCLSA-N
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoate
12879025
Crystal-Powder at 20°C
N-(tert-Butoxycarbonyl)-O-benzyl-L-serine N-Succinimidyl Ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator