missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440580050

 View more versions of this product

Catalog No. AC440580050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide
Authentic
248°C
34°C to 37°C
87694-52-8
100.0
C12H24N2O4
RRBFCGUIFHFYQK-VIFPVBQESA-N
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-3-methyl-1-oxobutan-2-yl]carbamate
10083961
Low Melting Solid or Liquid
113°C
-15
White to Yellow
5g
96.0
96% min. (GC)
2s-2-tert-butoxy carbonylamino-n-methoxy-3-methyl-n-methylbutanamide, boc-val-n och3 ch3, n-alpha-t-butyloxycarbonyl-n-methyl-n-methoxy-l-valine amide, n-boc-l-valine n'-methoxy-n'-methylamide, n-boc-l-valinen'-methoxy-n'-methylamide, n-tert-butoxycarbonyl-l-valine n'-methoxy-n'-methylamide, s-2-boc-amino-n-methoxy-n,3-dimethylbutanamide, tert-butyl n-1s-1-methoxy methyl carbamoyl-2-methylpropyl carbamate, tert-butyl s-1-methoxy methyl amino-3-methyl-1-oxobutan-2-yl carbamate
CC(C)C(C(=O)N(C)OC)NC(=O)OC(C)(C)C
260.34
260.34
97%

Chemical Identifiers

87694-52-8
260.34
2s-2-tert-butoxy carbonylamino-n-methoxy-3-methyl-n-methylbutanamide, boc-val-n och3 ch3, n-alpha-t-butyloxycarbonyl-n-methyl-n-methoxy-l-valine amide, n-boc-l-valine n'-methoxy-n'-methylamide, n-boc-l-valinen'-methoxy-n'-methylamide, n-tert-butoxycarbonyl-l-valine n'-methoxy-n'-methylamide, s-2-boc-amino-n-methoxy-n,3-dimethylbutanamide, tert-butyl n-1s-1-methoxy methyl carbamoyl-2-methylpropyl carbamate, tert-butyl s-1-methoxy methyl amino-3-methyl-1-oxobutan-2-yl carbamate
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-3-methyl-1-oxobutan-2-yl]carbamate
C12H24N2O4
RRBFCGUIFHFYQK-VIFPVBQESA-N
10083961
CC(C)C(C(=O)N(C)OC)NC(=O)OC(C)(C)C
SDS
Documents