missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-L-prolinamide, TCI America™

$96.25 - $337.64

Chemical Identifiers

CAS 35150-07-3
Molecular Formula C10H18N2O3
Molecular Weight (g/mol) 214.265
MDL Number MFCD00190827
InChI Key PITJAAIPVBVRAO-ZETCQYMHSA-N
Synonym boc-pro-nh2, 1-boc-l-prolinamide, boc-l-proline amide, l-1-n-boc-prolidinamide, s-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, boc-l-prolinamide, boc-l-pro-nh2, n-tboc-l-prolinamide, r-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, n-boc-l-prolinamide
PubChem CID 7019053
IUPAC Name tert-butyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37561G View Documents TCI America
B37561G
1g Each for $96.25
 
B37565G View Documents TCI America
B37565G
5g Each for $337.64
 
Specifications

Chemical Identifiers

35150-07-3
214.265
PITJAAIPVBVRAO-ZETCQYMHSA-N
7019053
CC(C)(C)OC(=O)N1CCCC1C(=O)N
C10H18N2O3
MFCD00190827
boc-pro-nh2, 1-boc-l-prolinamide, boc-l-proline amide, l-1-n-boc-prolidinamide, s-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, boc-l-prolinamide, boc-l-pro-nh2, n-tboc-l-prolinamide, r-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, n-boc-l-prolinamide
tert-butyl (2S)-2-carbamoylpyrrolidine-1-carboxylate

Specifications

White
1g
C10H18N2O3
boc-pro-nh2, 1-boc-l-prolinamide, boc-l-proline amide, l-1-n-boc-prolidinamide, s-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, boc-l-prolinamide, boc-l-pro-nh2, n-tboc-l-prolinamide, r-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, n-boc-l-prolinamide
CC(C)(C)OC(=O)N1CCCC1C(=O)N
214.265
214.27
≥98.0% (HPLC,N)
107°C
35150-07-3
MFCD00190827
PITJAAIPVBVRAO-ZETCQYMHSA-N
tert-butyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
7019053
Crystal-Powder at 20°C
N-(tert-Butoxycarbonyl)-L-prolinamide
Safety and Handling

Safety and Handling

TSCA : No