missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-L-norvaline, TCI America™

$19.86 - $55.00

Chemical Identifiers

CAS 53308-95-5
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD00037268
InChI Key INWOAUUPYIXDHN-ZETCQYMHSA-N
Synonym boc-l-norvaline, boc-nva-oh, n-boc-l-norvaline, boc-norvaline, 2s-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-2-aminovaleric acid, n-1,1-dimethylethoxy carbonyl-l-methionine, n-tert-butoxycarbonyl-l-norvaline, s-2-tert-butoxycarbonyl amino pentanoic acid, 2s-2-tert-butoxy carbonylamino pentanoic acid
PubChem CID 2733748
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CCCC(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B43201G View Documents TCI America
B43201G
1g Each for $19.86
 
B43205G View Documents TCI America
B43205G
5g Each for $55.00
 
Specifications

Chemical Identifiers

53308-95-5
217.265
INWOAUUPYIXDHN-ZETCQYMHSA-N
2733748
CCCC(C(=O)O)NC(=O)OC(C)(C)C
C10H19NO4
MFCD00037268
boc-l-norvaline, boc-nva-oh, n-boc-l-norvaline, boc-norvaline, 2s-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-2-aminovaleric acid, n-1,1-dimethylethoxy carbonyl-l-methionine, n-tert-butoxycarbonyl-l-norvaline, s-2-tert-butoxycarbonyl amino pentanoic acid, 2s-2-tert-butoxy carbonylamino pentanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid

Specifications

White
1g
C10H19NO4
boc-l-norvaline, boc-nva-oh, n-boc-l-norvaline, boc-norvaline, 2s-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-2-aminovaleric acid, n-1,1-dimethylethoxy carbonyl-l-methionine, n-tert-butoxycarbonyl-l-norvaline, s-2-tert-butoxycarbonyl amino pentanoic acid, 2s-2-tert-butoxy carbonylamino pentanoic acid
CCCC(C(=O)O)NC(=O)OC(C)(C)C
217.265
217.27
≥98.0% (T)
68°C
53308-95-5
MFCD00037268
INWOAUUPYIXDHN-ZETCQYMHSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
2733748
Crystal-Powder at 20°C
N-(tert-Butoxycarbonyl)-L-norvaline
Safety and Handling

Safety and Handling

TSCA : No