Learn More
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439430010
Specifications
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide | |
Glass Bottle | |
White | |
1g | |
97.5 | |
97.5% min. (GC) | |
2s-2-tert-butoxy carbonylamino-n-methoxy-n-methylpropanamide, boc-ala-n och3 ch3, boc-ala-nme ome, n-tert-butoxycarbonyl-l-alanine n'-methoxy-n'-methylamide, n2-tert-butoxycarbonyl-n-methoxy-n-methyl-l-alaninamide, r-tert-butyl1-methoxy methyl amino-1-oxopropan-2-ylcarbamate, s-2-boc-amino-n-methoxy-n-methylpropanamide, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-yl carbamate, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-ylcarbamate, tert-butyl n-1s-1-methoxy methyl carbamoyl ethyl carbamate | |
CC(C(=O)N(C)OC)NC(=O)OC(C)(C)C | |
232.28 | |
232.28 | |
98% |
Authentic | |
-25.5 | |
143.5°C to 148.3°C | |
87694-49-3 | |
100.0 | |
C10H20N2O4 | |
PWQIGBOSLQHOBT-ZETCQYMHSA-N | |
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-1-oxopropan-2-yl]carbamate | |
11806530 | |
Crystalline Powder or Powder |
Chemical Identifiers
87694-49-3 | |
232.28 | |
2s-2-tert-butoxy carbonylamino-n-methoxy-n-methylpropanamide, boc-ala-n och3 ch3, boc-ala-nme ome, n-tert-butoxycarbonyl-l-alanine n'-methoxy-n'-methylamide, n2-tert-butoxycarbonyl-n-methoxy-n-methyl-l-alaninamide, r-tert-butyl1-methoxy methyl amino-1-oxopropan-2-ylcarbamate, s-2-boc-amino-n-methoxy-n-methylpropanamide, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-yl carbamate, s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-ylcarbamate, tert-butyl n-1s-1-methoxy methyl carbamoyl ethyl carbamate | |
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-1-oxopropan-2-yl]carbamate |
C10H20N2O4 | |
PWQIGBOSLQHOBT-ZETCQYMHSA-N | |
11806530 | |
CC(C(=O)N(C)OC)NC(=O)OC(C)(C)C |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning